Anti-DDX58 Mouse Monoclonal Antibody (Biotin) [clone: Alme-1]

Supplier: Prosci
36-103
75792-874EA 861.6 USD
75792-874
Anti-DDX58 Mouse Monoclonal Antibody (Biotin) [clone: Alme-1]
Antibodies
RIG-I and MDA5 are highly conserved helicases involved in the innate immune response to virus. RIG-I is a member of the DEAD-box RNA helicases and is activated by cytoplasmic dsRNA and 5’-ppp RNA produced during the viral replication. The protein is characterized by a N-terminal region with two caspase recruitment domains (CARD) and a C-terminal region harboring potential ATP-dependent RNA helicase activity. RIG-I recruits the CARD adaptor inducing IFN-beta (Cardif) in a CARD-CARD-dependent manner resulting in NF-kappaB and IRF3 activation.

Immunohistochemistry: (paraffin sections). Immunoprecipitation: (1:200). Western blot: (1:1,000).

Type: Primary
Antigen: DDX58
Clonality: Monoclonal
Clone: Alme-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR