Stericheck Closed Sterility Test System, Type PFS, MDI
Supplier: MDI MEMBRANE TECHNOLOGIES, INC.
SPFV0902XXX202
75792-064CS 1685.21 USD
75792-064
Stericheck Closed Sterility Test System, Type PFS, MDI
Sterilizers Loop Sterilizers
Stericheck is a presterilized, nontoxic, non-pyrogenic, and ready to use sterility testing device designed for closed loop sterility testing of syringe needles as well products in the pre-filled syringes. The PFS series is specifically designed for use with pre-filled syringes.
  • Non-coring needle design
  • Pre-installed color coded clamps
  • Raised transparent vent
  • Customized needle connections
  • Validated for equal sample distribution
  • Compatible with Steripump or other available sterility testing equipment
The PFS system includes a specially designed adaptor for sterility testing of the syringe needle as well as its product
ETO Sterilized models can be used under a laminar flow hood. Gamma Sterilized models can be used in isolators or laminar flow hoods. PVDF Membrane models can be used for liquids with inhibitory properties, such as antibiotics. CN Membrane models can be used for all types of pharmaceutical products.
Testing of both inside and outside of the needle is ensured by liquid flow pattern. Raised vents prevent air flow obstruction due to fluid logging, and 'L' shaped flow directors prevent frothing. A tough membrane helps withstand inadvertent back pressure. Archivable sterilization indicator included in each pack.
Certifications: Validated as per USP 71 Sterility Tests. Validation Guide available on request.