Stericheck Closed Sterility Test System, Type SVP4, MDI
Supplier: MDI MEMBRANE TECHNOLOGIES, INC.
SV4V0902XXX202
75792-048CS 1685.21 USD
75792-048
Stericheck Closed Sterility Test System, Type SVP4, MDI
Sterilizers Loop Sterilizers
Stericheck is a presterilized, nontoxic, non-pyrogenic, and ready to use sterility testing device designed for closed loop sterility testing of Small Volume Parenterals (SVP). The SVP4 series is specifically designed for use with mini-vials and cartridges.
  • Non-coring needle design
  • Pre-installed color coded clamps
  • Raised transparent vent
  • Customized needle connections
  • Validated for equal sample distribution
  • Compatible with Steripump or other available sterility testing equipment
System includes a single needle with single tubing for transfer of sterile sample to a sterile pooling bottle, and a double vented needle with twin-tubing for transfer of pooled sample
ETO Sterilized models can be used under a laminar flow hood. Gamma Sterilized models can be used in isolators or laminar flow hoods. PVDF Membrane models can be used for liquids with inhibitory properties, such as antibiotics. CN Membrane models can be used for all types of pharmaceutical products.
Raised vents prevent air flow obstruction due to fluid logging, and 'L' shaped flow directors prevent frothing. A tough membrane helps withstand inadvertent back pressure. Archivable sterilization indicator included in each pack.
Certifications: Validated as per USP 71 Sterility Tests. Validation Guide available on request.