Puritan® PurFlock Ultra DNA-Free Standard Flock Swab with Tube, Puritan Medical Products
Supplier: Puritan Medical Products
2515061PFTTMC
75788-578BX 90.44 USD
75788-578
Puritan® PurFlock Ultra DNA-Free Standard Flock Swab with Tube, Puritan Medical Products
Swabs Swabs for Clinical Use
DNA-Free Regular Tip Applicator with Polystyrene Handle in Transport Tube. Ideal for DNA Collection.
  • Medical grade quality
  • Absorbent and soft smooth polyester flock tip
  • Tested to be DNA-free & RNase-free
  • Swab individually packaged in a transport tube (TT)
Ideal for general specimen collection for testing and screenig, as well as pediatric, opthalmological, and oropharyngeal applications. Can be used in veterinary care, wildlife forensic biology, DNA testing, and buccal or other cell collection.
Bonded securely to a hollow polystyrene handle that snaps when bent to a 90° angle. Comes packed in a resealable dry transport device for ease in transport to the lab. The (MC) style tube has a molded base that holds the swab tip in the center of the tube to help avoid loss of sample material during transport.
An ideal choice in flock swabs with excellent absorption and elution of specimens when HydraFlock's high levels of absorbency aren't necessary. Tested by an independent lab to the most stringent standards (less than 25 pg of human DNA).