PHCbi VIP® Series –86 °C Ultra-Low Temperature Chest Freezers, PHC Corporation
Supplier: PHC Corporation
MDF-DC700VXC-PA
10046-902EA 11845.21 USD
10046-902 75788-512
PHCbi VIP® Series –86 °C Ultra-Low Temperature Chest Freezers, PHC Corporation
Freezers Ultra Low Temperature Freezers
PHCbi's VIP® Series ultra-low temperature chest freezers are designed for safe, reliable storage of the most critical biologicals.
  • Reliable
  • Maximum storage
  • Minimal footprint
  • VIP Plus insulation composite
  • Uniform preservation
  • Microprocessor controls and alarms
  • Five-year manufacturer's parts and labor warranty
Energy efficiency in VIP® Series freezers is achieved through balanced temperature performance and power management
The robust refrigeration system is powered by PHCbi-engineered compressors. The system provides dependable cooling by balancing temperature performance and energy management. Reserve refrigeration power assures exceptional temperature recovery following door openings. VIP® freezers have demonstrated >99% reliability with excellent performance history.
The advanced combination of cabinet design, electronics, refrigeration, and critical components are engineered for enhanced security, performance, and reliability.
Ordering information: All units are supplied with a five-year manufacturer's parts and labor warranty.