Steri-Drape™ Urology Drape, 3M™
Supplier: Healthcare Products
MMM 1071
75787-234CS 897.08 USD
75787-234
Steri-Drape™ Urology Drape, 3M™
First Aid Supplies Surgical Drapes
Plastic specialty drape features an aperture and a built-in finger cot for use in urology and gynecology surgical procedures.