Cavilon™ Durable Barrier Cream, 3M™
Supplier: Healthcare Products
MMM 3355
75786-412CS 182.85 USD
75786-412
Cavilon™ Durable Barrier Cream, 3M™
Skincare Lotions Hand Creams
A concentrated cream that provides durable, long-lasting protection from body fluids while moisturizing the skin
  • Hypoallergenic
  • CHG compatible
  • Contains Dimethicone for skin protection
  • Fragrance-free
Unique, polymer-based formula provides a breathable barrier to promote skin health in patients at risk for skin damage from incontinence. The cream vanishes into the skin, does not clog briefs or diapers, and resists wash off. Using Cavilon Durable Barrier Cream regularly on intact skin can help prevent breakdown caused by regular exposure to urine and stool.