Transferrin ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT143-0001
75786-346EA 749.92 USD
75786-346
Transferrin ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive Transferrin ELISA kit enabling detection of transferrin in serum and plasma in just 2 hours.
  • Sensitive measurement of transferrin, detecting as little as 4.6 ng/ml
  • Negligible cross reactivity with similar proteins
  • High throughput format with results in 2 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of transferrin in human serum, human plasma and canine serum.
The Transferrin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Tracer, Conjugate, Substrate, and Stop Solution.