LH ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT107-0001
75786-328EA 322.64 USD
75786-328
LH ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive LH ELISA kit enabling detection of LH in serum, plasma and tissue culture media in just 2 hours.
  • Sensitive measurement of LH, detecting as little as 5.2 mIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in two hours for up to 39 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of LH in tissue culture media, serum and plasma in human and rat samples.
The LH ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in two hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution.