NGAL (Monkey) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BPD-KIT-045
75786-322EA 1995.79 USD
75786-322
NGAL (Monkey) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of NGAL in monkey urine, plasma, serum, tissue extracts, or culture supernatants.
  • Ready-to-use: Pre-coated plate and individual calibrators save time and reduce errors
  • Easy-to-use: Convenient storage at 4 °C with room temperature incubations
  • Rapid: Results in just four hours from up to 40 samples in duplicate
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in pig samples. Absorbance is read at 450 nm.
Ordering information: Contains Microtiter Plate, Sample Diluent Conc., Monkey NGAL Calibrator Stock, Wash Solution Conc., Biotinylated Monkey-NGAL Antibody, HRP-Streptavidin, TMB Substrate, Stop Solution. Manufactured by BioPorto Diagnostics.