FastDNA™ Spin Kit for Feces, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Designed to quickly and efficiently isolate PCR-ready genomic DNA from fresh or frozen human and animal stool samples.
- Pack size: 50 preparations
- Harndess: Ceramic Spheres, Vickers 800; Silica Spheres, Knoop 515; Glass Bead, Knoop 515
- Specific Gravity: Ceramic Spheres, 4; Silica Spheres, 2.5; Glass Bead, 2.5
Host cells, bacteria, fungi, viruses, protists and other cells present in fecal samples are easily lysed within 40 seconds with any FastPrep® instrument. The FastDNA SPIN kit for Feces contains pre-filled 2 ml Lysing Matrix tubes for the homogenization step as well as all the reagents needed to extract DNA in a purification process eliminating PCR inhibitors.
The Fast DNA® SPIN Kit for Feces quickly and efficiently isolates PCR-ready genomic DNA from fresh or frozen human and animal stool samples.
Kit contents: Lysing matrix E 50×2.0 ml tubes, PLS solution 25 ml, MT buffer 8 ml, sodium phosphate buffer 100 ml, PPS 25 ml, binding matrix 66 ml, wash buffer #1 120 ml, wash buffer #2 12 ml, TES 20 ml, BBS gel loading dye 200 μl, SPIN filter tubes 50 each, catch tubes 50 each.
Reagent: Sodium phosphate buffer - pH range: 7.8 to 8.2
Reagent: MT buffer - pH Range: 5.8 to 6.2
Reagent: PPS protein precipitate solution - pH range: 5.8 to 6.2
Reagent: Binding matrix - pH range: 6.5 to 7.2
Caution: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department. If working with biological, toxic, hazardous or infectious samples; Hand protection: Latex, Nitrile or other plastic material gloves. Skin and body protection: Labcoat and sleves. Eye protection: Safety glasses with side-shields Hygiene measures: Handle in accordance with good industrial hygiene and safety practice.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...