FastRNA™, Spin Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
116020050
75784-584EA 626.42 USD
75784-584 75784-586
FastRNA™, Spin Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastRNA™ Spin Kit isolates both large and small RNA species without the use of phenol or reducing agents.
  • Spin Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures
  • Spin Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi, and algae
The FastRNA™ SPIN Kit for Microbes quickly and efficiently isolates high-quality, total RNA from tough-to-lyse bacterial cell cultures and other microbial sources in approximately 15 minutes using a zirconium silicate Lysing Matrix (0.1mm) to lyse bacteria and SPIN columns for the purification process.
The FastRNA™ SPIN Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi, and algae in approximately 15 minutes using a specialized Lysing Matrix (Yttria-stabilized zirconium oxide, 0.5mm) for cell lysis and SPIN columns for the purification process.