Blue Biopsy Pads, Jaece
Supplier: Jaece
BP-1001
75783-972PK 39.05 USD
75783-972
Blue Biopsy Pads, Jaece
Biopsy Pads
Blue Biopsy Pads allow fluids to transfer safely and thoroughly around the tissue while holding the specimen secure.
  • Made from uniform polyester reticulated open cell foam
  • Transfer fluids safely around the tissue
  • Reduced risk of tissue sticking during embedding
  • Verified for consistency
Blue Biopsy Pads are made from uniform polyester reticulated open cell foam, which allows fluids to transfer safely and thoroughly around the tissue while holding the specimen secure.
The thin 2 mm design helps eliminate pressure from being applied to the unfixed specimen, reducing the risk of tissue sticking to the pads during embedding.