Miriad Rapid HCV/HIV Antibody Test LAB+, MedMira
Supplier: MEDMIRA
815311005954
75780-216BX 1167.77 USD
75780-216
Miriad Rapid HCV/HIV Antibody Test LAB+, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid HCV/HIV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis C and HIV-1/2
Miriad HCV/HIV features a built-in reagent control line, works with whole blood, serum, and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
In three easy steps users can obtain multiple instant results on a single test cartridge.
Ordering information: Each box of Miriad HCV/HIV LAB+ Format includes 50 test kits, 50 sample prep tubes, 50 transfer pipettes, and 2 x 30mL bottles of Universal Buffer.