Staticide® Static Sentry, ACL Staticide
Supplier: ACL Staticide
2006
75780-062CS 324.76 USD
75780-062
Staticide® Static Sentry, ACL Staticide
Surface Treatments Antistatic Treatments
Long lasting topical anti-static spray is ideal for eliminating charge generation on both porous and non-porous materials
Staticide® Static Sentry can be used on a variey of surfaces, including laminate flooring, carpeting, cabinetry, work surfaces, paper, polycarbonate, acrylic, PET, and glass. Safe to use on plastic and fabrics. No tribocharging to prevent dust attraction.
The spray works instantly, and is ozone safe and humidity independent. Ideal for use in offices, laboratories, institutions, and manufacturing facilities.
Certifications: RoHS compliant. Meets MIL-B-81705 Federal Test Standard 101.