Staticide® Anti-Static Plastic and Glass Cleaner, ACL Staticide
Supplier: ACL Staticide
8670
75780-010CS 165.73 USD
75780-010
Staticide® Anti-Static Plastic and Glass Cleaner, ACL Staticide
Surface Treatments Antistatic Treatments
Staticide® Anti-Static Plastic and Glass Cleaner effectively cleans, polishes, and neutralizes charges during use
This foam cleaning is ideal to use on computer monitors and housings. It keeps observation windows on production equipment in pristine condition longer than other cleaners and does not contain CFC's, HCFC's, or 1,1,1,trichloroethane.
The cleaner removes dirt, grease, ink, fingerprints, and nicotine stains. Non-streaking formula will not smear, scratch, or abrade soft plastic surfaces. Ideal for use on stainless steel surfaces as well. Transparent surfaces are left spotless for improved optical clarity and distortion-free reading.