Staticide® Turbo Blast Precision Duster, ACL
Supplier: ACL Staticide
8640
75780-007CS 528.72 USD
75780-007
Staticide® Turbo Blast Precision Duster, ACL
Sprays
Staticide® Turbo Blast Precision Duster is an economical, high-precision duster that offers maximum blasting power for electronic cleaning applications.
Filtered to 0.2 microns, this duster provides exceptional purity, and rids surfaces of particulate contamination without leaving residuals. Engineered to remove dirt, dust, oxide particles, and other airborne particles instantly. It is also excellent in accelerating the drying time of solvents. Includes straw for even greater pinpoint blasting.