Corning® Cryogenic Vial Grippers, Corning
Supplier: Corning
432136
75779-848CS 80.44 USD
75779-848
Corning® Cryogenic Vial Grippers, Corning
Forceps
These cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials.
They enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen.