Precision® Low Pressure Drop Inlet Liner with Wool for Bruker/Varian GCs equipped with 1177 Inlets, Restek
Supplier: Restek
22422
75779-160PK 282.02 USD
75779-160
Precision® Low Pressure Drop Inlet Liner with Wool for Bruker/Varian GCs equipped with 1177 Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.