Cup Inlet Liner with Wool for PerkinElmer Auto SYS™ and Clarus GCs equipped with Split/Splitless Inlets, Restek
Supplier: Restek
20836-213
75778-886PK 429.88 USD
75778-886
Cup Inlet Liner with Wool for PerkinElmer Auto SYS™ and Clarus GCs equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.