Precision® Inlet Liner with Top Taper and Wool for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Supplier: Restek
21029
75778-544PK 232.52 USD
75778-544
Precision® Inlet Liner with Top Taper and Wool for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.