SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37882-0000
74950-313PK 97.59 USD
74950-313
SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Reagent Reservoirs
This versatile reservoir is ideal for use when filling multi-tube blocks or microwell plates.
  • Graduated markings
  • Suitable for use with all brands of multichannel pipettors
  • Autoclavable
It features a graduated single reagent reservoir with graduations at 10, 25, 40, and 50 mL; a reservoir with eight separate 5 mL reagent channels; and a reservoir with 12 separate 5 mL reagent channels. Each compartment of the multichannel reservoirs is numbered for reagent identification.