SP Bel-Art Spill Containment Tray with Grid, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F24676-0003
74950-156EA 151.32 USD
74950-156
SP Bel-Art Spill Containment Tray with Grid, Bel-Art Products, a part of SP
Spill Control Materials
Large volume benchtop containment device protects users by capturing accidental or hazardous spills.
  • Epoxy-coated steel wire support grid
  • Molded polypropylene tray
  • Excellent chemical resistance
  • Autoclavable
An epoxy-coated wire support grid suitable for holding numerous vessels or tube racks is set within the large, deep polypropylene molded tray. Tray and grid have excellent chemical resistance. Steam autoclavable at 121°C (250°F).