SP Bel-Art Stackable Petri Dish Incubation Tray, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18983-1000
74950-152EA 105.27 USD
74950-152
SP Bel-Art Stackable Petri Dish Incubation Tray, Bel-Art Products, a part of SP
Racks Incubator Racks
Incubation trays increase overall incubator capacity by holding five 100 mm diameter or fifteen 60 mm diameter Petri dishes.
  • Polypropylene construction
  • Stackable
  • Large label area for easy identification
  • May be disinfected with common laboratory disinfectants
  • Autoclaving not recommended
Each polypropylene tray includes two large label areas for critical sample identification by marker or label and holes in the tray bottom to facilitate air flow.
Not recommended above 100°C (212°F). Autoclaving may warp. Can be disinfected with 70% ethanol or common laboratory disinfectants.