SP Bel-Art Multipurpose Tray with Lid, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F16230-0000
74950-090EA 56.14 USD
74950-090
SP Bel-Art Multipurpose Tray with Lid, Polypropylene, Bel-Art Products, a part of SP
Trays/Dishes/Pots Laboratory Trays/Dishes
Rugged, shallow tray may be used for safely storing small items, holding small vials or bottles in an ice or water bath, or holding instruments while autoclaving.
  • Molded polypropylene construction
  • Stackable for efficient storage
  • Inside corners are smooth and rounded
  • Easy to clean
Inside tray corners are smooth and rounded for easy cleaning. Stacks with identical trays and lids for efficient storage. Both tray and lid are constructed of molded polypropylene. Steam autoclavable at 121°C (250°F).