Nalgene® 4-in-1 EZ Tote™ Labware Carrier, HDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
6565-0001
74910-028CS 259.83 USD
74910-028
Nalgene® 4-in-1 EZ Tote™ Labware Carrier, HDPE, Thermo Scientific
Carriers Bottle Carriers/Containers
Unique lightweight carrier enables safe and easy transport of bottles and other labware
Constructed of durable corrugated HDPE plastic, it holds up to 12.7kg (28lbs.) in the form of eight 500mL, six 1L, or two 4L bottles. Carrier stores flat and can be assembled quickly with preassembled partitions. Instructions are printed on each unit. Recommended for use in room temperature or refrigerated environments. Not autoclavable.