Nunc® Barcode Scanners
Supplier: Thermo Fisher Scientific
330050
73521-418CS 1122.37 USD
73521-418
Nunc® Barcode Scanners
Barcode and RFID Systems
Nunc bar code scanners with plug and play functionality play a critical role in sample tracking
The scanners automatically switch between code 128, code 39, interleave 2 of 5 and several other barcodes. These scanners are durable low-cost, and easy to use.