Nunc® Vertical Microplate Storage Rack, Thermo Scientific
Supplier: Thermo Fisher Scientific
367002
73521-338CS 299.34 USD
73521-338
Nunc® Vertical Microplate Storage Rack, Thermo Scientific
Racks Microplate Racks
Store and transport samples in microplates, deep-well plates, and cryobank racks safely and conveniently using Nunc® Vertical Microplate storage racks
These autoclavable aluminum racks are ideal for genome library storage, DNA and RNA libraries, and storage of compounds for high throughput screening.