Security-Snap™ Sterile Coliform Water Sample Bottle, Thermo Scientific
Supplier: Thermo Scientific
411G10R27
73520-380CS 438.78 USD
73520-380
Security-Snap™ Sterile Coliform Water Sample Bottle, Thermo Scientific
Bottles Water Sampling Bottles
The Security-Snap™ Sterile Coliform Water Sample Bottle meets all the requirements for microbiology sample containers as specified by the 'EPA Manual for the Certification of Laboratories Analyzing Drinking Water'.
Manufactured from non-toxic, non-autofluorescent polypropylene with a molded-in 100±2.5 ml fill line.
The bottle features a Security-Snap™ flip-top lid which provides tamper evidence before and after sample collection, while protecting against any possibility of accidental opening during transport.