Molecular BioProducts Solvent Safe Carbon Filtered Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
5079EB
72830-004CS 954.06 USD
72830-004 72830-014
Molecular BioProducts Solvent Safe Carbon Filtered Pipette Tips, Thermo Scientific
Pipette Tips
MicroPoint design for accurate low-volume pipetting.
  • Volume: 100 to 1000 µl
  • Non-sterile
  • Certified RNase-, DNase-, DNA- and pyrogen-free
  • Bulk pack
Tips, pipette, filter, 200 µl, MicroPoint, with reference marks, Manual pipetting of organic solvents, 500 tips per bag.
Solvent safe carbon filter protects pipettors from damage by blocking corrosive vapors from volatile organic solvents.