ART® Solvent Safe Micropoint Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
5479E
72830-008CS 1792.37 USD
72830-008 72830-010
ART® Solvent Safe Micropoint Pipette Tips, Thermo Scientific
Pipette Tips
ART specialty pipette tips are available in extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel, gel loading for loading acrylamide or agarose gels, wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials, and Solvent Safe for acids, bases and aggressive organic solvents.