Kjeldahl Distillation Unit, 2-Place, Labconco®
Supplier: Labconco
2128501
72430-070EA 7850 USD
72430-070
Kjeldahl Distillation Unit, 2-Place, Labconco®
Distillation Systems Kjeldahl Distillation Systems
Designed for mounting on a base cabinet or table, unit provides durability for laboratories which require a separate distillation station.
Frame is constructed of powder-coated steel. Unit features two 600-watt ceramic heaters for distillation, operated by two individual infinite-control knobs with variable input from 20 to 100% of capacity. Distillation manifold has two seamless stainless steel tubes with 3/8" NPT inlet and outlet water connections. Connection to water source of no less than 5/16" required.
Ordering information: Supplied with eight-foot, three-wire cord and plug.