Bunsen Burners, Humboldt
Supplier: Humboldt Manufacturing
H-5890
72410-060EA 46.35 USD
72410-060
Bunsen Burners, Humboldt
Heaters Gas Burners
Adjustable burners with threaded needle valves utilize the highquality die-cast bases and provide a precision needle valve to achieve an accurate, adjustable flame.
Overall Height: 156 mm
Mixing Tube OD: 13 mm