Standard Metal Hub Luer Lock Needles
Supplier: Hamilton
90018 90028 90014 90013 90012 90032 90029 90011 90025 90023 90038 90021 90010 90053 90020 90027 7730-04 7729-09 7729-07 7729-06 7729-04 7729-01 91022 90017 90016 90015 90019 91012 7729-02 7729-03 7729-08 7729-10 7729-11 7729-12 7729-13 7729-14 7730-01 7730-02 7730-05 7748-08 7749-03 90051 90052 91016 7729-05 91010 91011 91013 91014 91015 91017 91018 91019 7730-03 91028 91029 91030 91031 91032 91033
60377-969PK 79.2 USD
60377-969 60378-065 72310-208 72310-218 72310-238 72310-240 72310-242 72310-244 72310-246 72310-248 72310-250 72310-252 72310-268 72310-270 74980-010 74980-012 82010-348 82010-360 82010-364 82010-366 82010-370 82010-376 82010-460 72310-160 72310-180 72310-200 72310-266 89210-104 89220-896 89220-898 89220-902 89220-904 89220-906 89220-908 89220-910 89220-912 89220-914 89220-916 89220-918 89220-948 89220-968 89221-156 89221-158 89221-546 89220-900 10217-404 10217-406 10217-408 10217-410 10217-412 10217-414 10217-416 10217-418 10217-420 10217-430 10217-432 10217-434 10217-436 10217-438 10217-440
Standard Metal Hub Luer Lock Needles
Syringe Needles
Hamilton company offers the most comprehensive selection of standard and custom needles in the industry.
  • Needles are made of 304 stainless steel
Each is manufactured to achieve the highest level of accuracy and precision possible. Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors.
PTFE Luer Lock (TLL)- This termination has a PTFE, male luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters and dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 ml and greater syringes. Repeated autoclaving will shorten syringe life.
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34 to 10. Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34 to 10. Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26 to 10.