MULTI-AD® Fluid Dispensing System, B. Braun
Supplier: B Braun
513501
72220-000CS 634.02 USD
72220-000
MULTI-AD® Fluid Dispensing System, B. Braun
Catheters
Transfer set with weighted sinker and tubing clip for use with MULTI-AD® pump or for manual activation use when dispensing fluids from an open container
The set is DEHP-free and latex-free. Includes a 10cc luer lock syringe with adjustable automatic spring return and dual check valve.