Paraffin wax blended with synthetic polymers, Solidification point: 55-57°C type 1 for infiltration
Supplier: Epredia
Find the perfect paraffin blend to fit your needs with Epredia™ Paraffin Type 1, 3, 6, 9: Four different blends of highly refined paraffin and polymers, offering a choice of infiltration times and cutting characteristics.
- All paraffins melt between 55° and 57°C and are triple filtered to 0.5µm
- Flake form has a greater surface-to-weight ratio than pellets
- Faster melting time
- Type 1 contains no polymer and offers rapid infiltration
- Type 3 contains minimal polymer and offers fast infiltration
- Type 6 is harder than Type 3 and allows for thinner sections
- Type 9 has the highest polymer content available and yields extremely thin sections
These blends of highly refined paraffin and polymers offer a choice of infiltration times and cutting characteristics to meet the needs of individual laboratories. They feature excellent ribboning properties, melting points of 55 to 57°C (131 to 135°F), and triple filtration to 0.5µm. Case of ten 0.9kg (2lbs.) bags.
Type 6: Higher polymer content than Type 3. Allows thinner sections.
Type 9: Highest polymer content. Yields ultra-thin sections.
CAS Number:
8002-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...