Camco® Sequester-Sol® Liquid Anticoagulant, Cambridge
Supplier: CAMBRIDGE DIAGNOSTIC PRODUCT
205
71140-002EA 80.21 USD
71140-002
Camco® Sequester-Sol® Liquid Anticoagulant, Cambridge
Clinical Diagnostic Supplies Clinical Reagents and Kits
This effective, nontoxic anticoagulant is used in hematology and blood chemistry procedures.
It stabilizes the cellular constituents of blood in the same manner as lavender top vacuum tubes. Complete blood counts, as well as blood smears for staining, can be obtained within 48 hours after collection of the specimen without any distortion of the cellular constituents.
Contains an aqueous solution of dipotassium EDTA.
Directions for use: Place one drop of solution into a clean blood collection tube. Then add 5 ml of venous blood and invert tube 4 to 5 times.
Packaging: Packaged in a polyethylene bottle with a dropper tip for accurate dispensing. Size: 29.5 ml (1 oz.).