Wright-Giemsa stain, Camco® Quik Stain® II
Supplier: CAMBRIDGE DIAGNOSTIC PRODUCT
Synonyms: Wright-Giemsa Solution
211
71130-004EA 173.88 USD
71130-004
Wright-Giemsa stain, Camco® Quik Stain® II
Wright-Giemsa stain (solutions & kits)
The Fastest and Most Reliable Differential Stain Available
  • Fast: Requires less than 30 seconds
  • Reliable: Uniformity from slide to slide
  • Economical: Stains hundreds of smears
  • Simple to Use: Only one staining solution necessary
Camco Quik Stain revolutionized differential staining. One easy to use solution is all that is required for manual staining techniques. Both products incorporate a buffer into the stain solution eliminating many additional steps. Provides quick differential staining of blood smears in just 30 seconds. These products are great for a backup system in your lab or when a STAT result is necessary. These stains should be used with distilled water (pH 6-7) as the rinse.
Directions:
Prepare blood smear on slide.
Dip slide in stain for 10 seconds.
Dip slide in distilled water (pH 6-7) for 20 seconds (or more for darker staining).
Air dry.
Caution: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm. For more information, go to P65Warnings.ca.gov.