Silver nitrate, crystals ACS

Supplier: VWR
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

ANAC79626-140
CA71008-410EA 282.73 USD
CA71008-410 71008-412 71008-418
Silver nitrate, crystals ACS
Silver nitrate

Prices subjet to market values

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (AgNO3) 99.0% min.
Clarity of solution Passes Test
Chloride (Cl) 5 ppm max.
Free acid Passes Test
Substances not precipitated by HCl 0.01% max.
Sulfate (SO4) 0.002% max.
Copper (Cu) 2 ppm max.
Iron (Fe) 2 ppm max.
Lead (Pb) 0.001% max.
Appearance: Colourless, large crystals or white small crystals info only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR