Silver ≥99.99%, foil 0.500 x 0.005 in

Supplier: REFINING SYSTEMS

AG-0.500X0.005IN-5N
71008-244EA 267.05 USD
71008-244
Silver ≥99.99%, foil 0.500 x 0.005 in
Silver

Refining Systems, Inc. manufactures foils in different widths and thickness in pure or alloy form with different materials.


Gold/Palladium and Platinum/Palladium foils can be manufactured as well.


Ordering information: Contact your VWR sales representative for more information.

Formula: Ag
MW: 107.87 g/mol
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR