Hydrofluoric acid 47 - 51%, Environmental Grade Plus

Supplier: SEASTAR CHEMICALS
Danger

S020502-ANNF07 46564-320
CA71007-536EA 4627.02 USD
CA71007-536 71007-532
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Hydrofluoric acid
Formula: HF
MW: 20.01
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 II

Order Now

Specification Test Results

Assay 47 - 51%
Aluminum (Al) 20 ppt max.
Antimony (Sb) 20 ppt max.
Arsenic (As) 50 ppt max.
Barium (Ba) 10 ppt max.
Beryllium (Be) 10 ppt max.
Bismuth (Bi) 10 ppt max.
Boron (B) 100 ppt max.
Cadmium (Cd) 10 ppt max.
Calcium (Ca) 10 ppt max.
Cerium (Ce) 10 ppt max.
Cesium (Cs) 10 ppt max.
Chromium (Cr) 10 ppt max.
Cobalt (Co) 10 ppt max.
Copper (Cu) 10 ppt max.
Dysprosium (Dy) 1 ppt max.
Erbium (Er) 1 ppt max.
Europium (Eu) 1 ppt max.
Gadolinium (Gd) 1 ppt max.
Gallium (Ga) 10 ppt max.
Germanium(Ge) 10 ppt max.
Gold(Au) 20 ppt max.
Hafnium (Hf) 10 ppt max.
Holmium (Ho) 1 ppt max.
Indium (In) 1 ppt max.
Iron (Fe) 10 ppt max.
Lanthanum (La) 10 ppt max.
Lead (Pb) 10 ppt max.
Lithium (Li) 10 ppt max.
Lutetium (Lu) 1 ppt max.
Magnesium (Mg) 10 ppt max.
Manganese (Mn) 10 ppt max.
Mercury (Hg) 50 ppt max.
Molybdenum (Mo) 10 ppt max.
Neodymium (Nd) 1 ppt max.
Nickel (Ni) 20 ppt max.
Niobium (Nb) 10 ppt max.
Palladium (Pd) 20 ppt max.
Platinum (Pt) 20 ppt max.
Potassium (K) 10 ppt max.
Praseodymium (Pr) 1 ppt max.
Rhenium (Re) 10 ppt max.
Rhodium(Rh) 20 ppt max.
Rubidium (Rb) 20 ppt max.
Ruthenium (Ru) 20 ppt max.
Samarium (Sm) 1 ppt max.
Scandium (Sc) 10 ppt max.
Selenium (Se) info only
Silver (Ag) 10 ppt max.
Sodium (Na) 10 ppt max.
Strontium (Sr) 10 ppt max.
Tantalum (Ta) info only
Tellurium (Te) 1 ppt max.
Terbium (Tb) 1 ppt max.
Thallium (Tl) 10 ppt max.
Thorium (Th) 1 ppt max.
Thulium (Tm) 1 ppt max.
Tin (Sn) 20 ppt max.
Titanium (Ti) 20 ppt max.
Tungsten (W) 20 ppt max.
Uranium (U) 1 ppt max.
Vanadium (V) 10 ppt max.
Ytterbium (Yb) 1 ppt max.
Yttrium (Y) 1 ppt max.
Zinc (Zn) 10 ppt max.
Zirconium (Zr) 10 ppt max.
Total sulfur 100 ppt max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR