Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ethyl alcohol denatured

ANAC38344-540
71007-230CS 1546.88 USD
71007-230
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Ethanol denatured

(DAG-2A)

Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1986
ADR: 3 (6.1),II

Order Now

Specification Test Results

E/E 80.2% v/v min.
Appearance (clear, colorless liquid) Passes test
Colour (APHA) 5 max.
Density at 20°C 0.8106 g/ml max.
Acidity (as CH3COOH ) 0.003% w/v max.
Ethyl acetate 0.75 - 0.95% v/v
Methanol 12.95 - 14.45% v/v

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR