Ammonia solution 28% ACS

Supplier: Anachemia
Danger

05566-461 ANAC05566-460
CA71006-578CS 513.69 USD
CA71006-578 71006-576
Ammonia solution 28% ACS
Ammonia solution
Formula: NH₃
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance: Colourless and free from suspended matter or sediment Conforms
Assay (as NH3) 28.0 - 30.0 %
Residue after ignition 0.002% max.
Carbon Dioxide (CO2) 0.002% max.
Chloride (Cl) 0.5 ppm max.
Phosphate (PO4) 2 ppm max.
Nitrate (NO3) 2 ppm max.
Sulfate (SO4) 2 ppm max.
Heavy metals (as Pb) 0.5 ppm max.
Iron (Fe) 0.2 ppm max.
Substances reducing KMnO4 Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR