VWR® Antibody Signal Enhancer
Supplier: VWR
M336-1L
71005-960EA
756.3
USD
71005-960
71005-964
VWR® Antibody Signal Enhancer
Protein Linking and Labeling Systems
Antibody Signal Enhancer is a Western blotting antibody diluent that increases intensity of protein detection up to 5-fold compared to conventional blotting. The enhancer is a ready-to-use solution that replaces TBS-T with non-fat dry milk, or other diluent, for antibody incubation steps. It is formulated for use with HRP- and biotin-conjugated antibodies and works with both PVDF and nitrocellulose blots.
- Improve detection of low abundance proteins
- Conserve valuable antibody reagents
- No extra incubation steps required
Antibody Signal Enhancer is stable at room temperature and is a non-hazardous formulation.