Water, AquaPur™ ST USP, purified, sterile

Supplier: Decon Labs

8904
71003-690CS 327.79 USD
71003-690
Water, AquaPur™ ST USP, purified, sterile
Water

AquaPur Purified Water has been tested to meet USP/EP specifications for purified water. Packaged in convenient point of use packaging for laboratory or production applications requiring purified water. The product is filtered to 0.2 µm and is guaranteed to meet specifications through expiration date.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

USP Water Conductivity USP 645 1.3 umhos/cm At the time of manufacturing
USP Total Organic Carbon USP 643 0.5 ppm
Endotoxin Testing 0.25 EU/mL
USP Sterility testing Pass Filter Membrane Test (14 day incubation)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR