Ethanol denatured absolute (200 Proof)

Supplier: Decon Labs

29043A
71003-544CS 301.24 USD
71003-544
Ethanol denatured absolute (200 Proof)
Ethanol denatured

Based on SDA 3A formula.


Ordering information: There is not tax on denatured ethanol, but users are still required to have the Industrial Alcohol User Permit, marked for 'Withdraw and use of specially denatured alcohol/rum'.

Formula: H₃CCH₂OH
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR