Iodoacetamide
Supplier: G-Biosciences
|
Warning
|
G-Biosciences offers proteomic grade iodoacetamide in two bulk quantities (5 or 25 grams) or in OneQuant™ format.
G-Biosciences' OneQuant™ Iodoacetamide are single aliquots of iodoacetamide that eliminate the need for weighing, preventing loss of reagent
Formula:
ICH₂CONH₂ Boiling Pt: 266 °C (1013 hPa) Melting Pt: 91…94 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008028 CAS Number: 144-48-9 EINECS: 205-630-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...