Guanidinium thiocyanate for biochemistry

Supplier: G-Biosciences
Warning

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

RC-058 RC-059
71003-412EA 113.42 USD
71003-412 71003-414
Guanidinium thiocyanate for biochemistry
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR