Glycine for biochemistry
Supplier: G-Biosciences
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
RC-055
RC-054
71003-408EA
113.21
USD
71003-408
71003-406
Glycine for biochemistry
Glycine
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: 83 °C Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...