3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) 5% in aqueous solution

Supplier: G-Biosciences

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS

DG097
71003-310EA 214.87 USD
71003-310
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) 5% in aqueous solution
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR